Nunc™ Rectangular Dishes
Nunc™ Rectangular Dishes
Nunc™ Rectangular Dishes
Thermo Scientific™

Nunc™ Rectangular Dishes

Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
2670628
2670601
2670614
Catalog number 267062
Price (MXN)
-
No. of Wells:
8
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.

Rectangular Dishes 1, 4 and 8-Well

  • 1-well dish is ideal for HTS Petri dish applications
  • 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
  • 4-well dish holds four glass slides for storage

1-Well Format

  • Convenient plate format for cell culture
  • HTS Petri dish applications

4- and 8-Well Formats

  • Assay compartmentalization
  • Probe multiple gene subsets
  • Wash Western blot strips
  • 4-well dish holds four glass slides
  • Useful in all areas of cell culture, including scale-up and cloning

Ordering Information:

Customized bar coding is available

Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatSBS Standard
Height (Metric)59 mm
IncludesLid
Length (Metric)100 mm
No. per Case100/Cs.
No. per Pack10 Pk.
Product TypeCell Culture Dish
Quantity10 Pk.
Total Volume (Metric) Well11 mL
Width (Metric)86 mm
Array2 x 4
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells8
SterilitySterile
Surface TreatmentUntreated
Well ShapeRectangular
Unit SizeCase of 100