Product Image
Thermo Scientific™
Nalgene™ Silicone Sanitary Gaskets (19 mm ferrule)
For Use With: Nalgene Carboys
Have Questions?
Catalog NumberQuantity
2672-0075Case of 6
Catalog number 2672-0075
Price (USD)
113.00
Case of 6
Quantity:
Case of 6
Price (USD)
113.00
Case of 6
Protect against leakage, breakage and contamination by using Thermo Scientific™ Nalgene™ Silicone Sanitary Gaskets for Nalgene™ Carboys. Constructed of platinum-cured silicone, the gaskets ensure the highest degree of purity for biopharmaceutical applications.
  • Autoclavable
  • Use with Nalgene™ Carboys (Cat. Nos. 2261, 2630, and 2640)
Specifications
Certifications/ComplianceMeets USP Class VI requirements
Closure SizeFerrule size 0.75 in. (19 mm)
DescriptionSilicone Sanitary Gaskets
For Use With (Equipment)Nalgene Sanitary Carboys
No. per Case6/Cs.
No. per Pack1 Ea.
QuantityCase of 6
AutoclavableAutoclavable
MaterialSilicone
Product TypeGasket
Unit SizeCase of 6