Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Thermo Scientific™
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
The Thermo Scientific™ Nunc™ Edge 2.0 Plate provides an efficient solution for preventing evaporation and in turn the risks associated with edge effect.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호케이스당 수
26742750 Microplates
267544160 Microplates
카탈로그 번호 267427
제품 가격(KRW)
177,000
온라인 행사
Ends: 31-Dec-2026
196,000
할인액 19,000 (10%)
Case of 50
케이스당 수:
50 Microplates
제품 가격(KRW)
177,000
온라인 행사
Ends: 31-Dec-2026
196,000
할인액 19,000 (10%)
Case of 50
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
사양
인증/적합성All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
배양 면적0.35 cm2/well
뚜껑With Lid
케이스당 수50 Microplates
팩당 수1
포장1/Pk., 50/Cs.
제품 유형Microplate
스커트 스타일Low Flange
부피(미터법) 웰400 μL
어레이8 x 12
색상Clear
물질Polystyrene
웰 수96
제품라인Nunc
멸균Sterile
표면 처리Untreated
웰 형상Flat Bottom (F-well)
Unit SizeCase of 50