Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Thermo Scientific™
Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Green features
Lower spending when storing compounds for small volume applications by using these 384-shallow well standard height polypropylene microplates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityFit for Purpose
267461BlackNon-sterileCertified Quality
267460NaturalSterileCertified Quality
267462WhiteNon-sterileCertified Quality
Catalog number 267461
Price (USD)
1,052.35
Online exclusive
1,228.15
Save 175.80
Case of 100
Color:
Black
Sterility:
Non-sterile
Fit for Purpose:
Certified Quality
Price (USD)
1,052.35
Online exclusive
1,228.15
Save 175.80
Case of 100
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
Features:
  • Rounded wells are optimized for mixing and sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Natural plates for storage identification or storing colorimetric assays
  • White plates for luminescent assays
  • Black plates for fluorescent assays
  • Customized bar coding is available
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
Dnase FreeYes
Description384-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
LidWithout Lid
No. per Case100
No. per Pack25
Packaging100 Cases
Plate SizeANSI/ SLAS Standard
Product Type384-well plate
Quantity100/Cs.
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well55 μL
Volume (Metric) Working2 to 35 μL
Array16 x 24
BarcodeNo Barcode
ColorBlack
Fit for PurposeCertified Quality
MaterialPolypropylene
No. of Wells384
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeSquare, Round Bottom (U-well)
Unit SizeCase of 100
Frequently asked questions (FAQs)
Do you offer Nunc Microplate Lids for the Nunc Black or White 384-Well Microplates?
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.
Why do you offer Nunc 384-Well Black or White Microplates when the cells cannot be observed using an inverted light microscope?
We offer Nunc Black or White 384-Well microplates that are intended for use in conjunction with 384-well microplate luminometers and fluorometers that have excitation and detection systems located above the plate. Due to the opaque quality of these microplates, all detection systems must rely on either luminescent or fluorescent reagents, such as viability stains that report cell activity and membrane integrity.