Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Thermo Scientific™
Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Have Questions?
Catalog NumberQuantity
267578160/Cs.
Catalog number 267578
Price (USD)
327.00
Case of 160
Quantity:
160/Cs.
Price (USD)
327.00
Case of 160
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Culture Area0.35 cm2/well
LidWith Lid
No. per Case160
No. per Pack10
Packaging10 Pk., 160 Cases
Product TypeMicroplate
Quantity160/Cs.
Skirt StyleLow Flange
Volume (Metric) Well400 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentUntreated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 160