Methanol, 99.8%, for HPLC
Methanol, 99.8%, for HPLC
Methanol, 99.8%, for HPLC
Thermo Scientific Chemicals

Methanol, 99.8%, for HPLC

Green features
For HPLC | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2682800101 LGlass Bottle
Catalog number 268280010
Price (USD)
66.70
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
66.70
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.8 %
UVat 260 nm A: =<0.005
Acidity/alkalinity=<0.0002 meq/g
Wavelength at 1 A.U.=<205 nm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only