Dichloromethane, 99.8%, for HPLC, stabilized with amylene
Dichloromethane, 99.8%, for HPLC, stabilized with amylene
Dichloromethane, 99.8%, for HPLC, stabilized with amylene
Thermo Scientific Chemicals

Dichloromethane, 99.8%, for HPLC, stabilized with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 268330025
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.8 %
Acidity=<0.0001 meq/g
UVat 233 nm A: =<1
UVat 250 nm A: =<0.02
UVat 280 nm A: =<0.005
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only