Lithium acetate, anhydrous, 99%, extra pure
Lithium acetate, anhydrous, 99%, extra pure
Lithium acetate, anhydrous, 99%, extra pure
Thermo Scientific Chemicals

Lithium acetate, anhydrous, 99%, extra pure

CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 268640010
Price (USD)
225.65
Online exclusive
250.00
Save 24.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
225.65
Online exclusive
250.00
Save 24.35
Each
Chemical Identifiers
CAS546-89-4
IUPAC Namelithium(1+) acetate
Molecular FormulaC2H3LiO2
InChI KeyXIXADJRWDQXREU-UHFFFAOYSA-M
SMILES[Li+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder
Melting point280°C to 285°C
Water=<0.5 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only