Nalgene™ Polycarbonate, End Cap
Thermo Scientific™
Nalgene™ Polycarbonate, End Cap
For Use With: Nalgene Carboy 2630 and 2261
Catalog NumberQuantity
2688-2075Case of 4
Catalog number 2688-2075
Price (JPY)
16,300
Case of 4
Quantity:
Case of 4
Sanitary ports for easy fill/ing and dispensing. Thermo Scientific™ Nalgene™ End Caps in polycarbonate or polypropylene to provide operations. Caps also can be securely mounted to 3 in. sanitary flange for applications used with Nalgene™ Sanitary Carboys.
  • Autoclavable
  • Compatible with Nalgene Sanitary Carboys (Cat. Nos. 2630 and 2261)
Specifications
DescriptionTwo 0.75 in. (19mm) sanitary ports, polycarbonate
Diameter (English) Inner Tubing0.75 in.
Diameter (Metric) Inner Tubing19 mm
For Use With (Equipment)Nalgene Carboy 2630 and 2261
No. per Case4/Cs.
No. per Pack1 Ea.
QuantityCase of 4
MaterialPolycarbonate
Product TypeEnd Cap
Unit SizeCase of 4