n-Butyl propionate, 99+%
n-Butyl propionate, 99+%
n-Butyl propionate, 99+%
Thermo Scientific Chemicals

n-Butyl propionate, 99+%

CAS: 590-01-2 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 269480010
Price (USD)
138.65
Special offer
Online exclusive
Ends: 28-Dec-2026
163.00
Save 24.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
138.65
Special offer
Online exclusive
Ends: 28-Dec-2026
163.00
Save 24.35
Each
Chemical Identifiers
CAS590-01-2
IUPAC Namebutyl propanoate
Molecular FormulaC7H14O2
InChI KeyBTMVHUNTONAYDX-UHFFFAOYSA-N
SMILESCCCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4000 to 1.4020 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only