Ethyl 3-(N,N-dimethylamino)acrylate, 99+%
Ethyl 3-(N,N-dimethylamino)acrylate, 99+%
Ethyl 3-(N,N-dimethylamino)acrylate, 99+%
Thermo Scientific Chemicals

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%

CAS: 924-99-2 | C7H13NO2 | 143.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 269630250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS924-99-2
IUPAC Nameethyl (2Z)-3-(dimethylamino)prop-2-enoate
Molecular FormulaC7H13NO2
InChI KeyMVUMJYQUKKUOHO-WAYWQWQTSA-N
SMILESCCOC(=O)C=C/N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light orange
Refractive index1.5105 to 1.5125 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only