Nunc™ MicroWell™ 96-Well Microplates
Thermo Scientific™
Nunc™ MicroWell™ 96-Well Microplates
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLidSkirt StyleBarcode
269787SterileWithout LidLow Flange PinchbarNo Barcode
260887SterileWith LidHigh FlangeIncludes Barcode
260844Non-sterileWithout LidHigh FlangeIncludes Barcode
Catalog number 269787
Price (MXN)
-
Sterility:
Sterile
Lid:
Without Lid
Skirt Style:
Low Flange Pinchbar
Barcode:
No Barcode
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
  • Made from clear polystyrene with excellent optical properties, ideal for optical measurements.
  • Non-treated plate with hydrophobic surface is suited for suspension cell cultures and a variety of biochemical assays.
  • Cell culture plate received the Thermo Scientific™ Nunclon™ Delta surface treatment that promotes cell adhesion and growth
  • Collagen I and Poly-D-Lysine coated plates are enhanced for finicky adherent cells and biomolecule attachment
  • Different flange designs are compatible with either manual or robotic operations
  • Customized bar coding is available
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Description96F well plate, Non-Treated, clear, without lid, Sterile (Pack of 1, Case of 50)
FormatStandard
LidWithout Lid
No. per Case50
No. per Pack1
Packaging1 Pk.
Product TypeMicroplate
Skirt StyleLow Flange Pinchbar
Array8 x 12
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50