dU Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

dU Phosphoramidite, standard grade, 20-400 finish bottle

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporationRead more
Have Questions?
Catalog NumberQuantity
27-1738-080.25 g
Catalog number 27-1738-08
Price (USD)
60.25
Each
Quantity:
0.25 g
Price (USD)
60.25
Each

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 30 mL amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C39H47N4O8P
• Base—dU
• Formula weight—730.8 g/mol
• CAS number—109389-30-2
• HPLC Purity—≥98.0%; conforms to reference
31P NMR Purity—≥98.0%; conforms to reference

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Expedite Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxyuridine (dU)
Product TypedU CEP
Purity98%
Quantity0.25 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).