TheraPure 2ʹ-OMe-iBu-G Phosphoramidite
Thermo Scientific™

TheraPure 2ʹ-OMe-iBu-G Phosphoramidite

2'-OMe-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
27-2046-041 gSerum Vial Bottle
27-2046-101 gAmber Bottle (20-400 Thread)
27-2046-75100 gBulk Standard
Catalog number 27-2046-04
Price (USD)
23.53
Each
Quantity:
1 g
Format:
Serum Vial Bottle
Price (USD)
23.53
Each

2'-OMe-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in an amber bottle.

Available with a Serum Vial, 20-400 Finish, or 28-400 Finish neck style.

  • Serum vial neck style is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.
  • 20-400 finish neck style is compatible with most Expedite synthesizers.
  • 28-400 finish neck style is compatible with most MerMade synthesizers.

Features:

  • Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methyl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula—C45H56N7O9P
  • Base—G
  • Base Protection—iBu
  • Formula weight—869.9 g/mol
  • CAS number—150780-67-9
  • HPLC purity—≥99%; M-11 Impurity: ≤0.01%; Single critical impurity ≤0.3%
  • 31P NMR purity—>99%; Sum of non-primary peaks at 140-152 ppm ≤0.5%
For Research Use Only.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
FormatSerum Vial Bottle
GradeTheraPure™
Nucleic Acid TypeRNA
Nucleoside TypeGuanosine (G)
Product Type2in-OMe-G CEP
Purity99%
Quantity1 g
Reagent Features2in-OMe modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).