Hydroxylamine hydrochloride, 99+%
Hydroxylamine hydrochloride, 99+%
Hydroxylamine hydrochloride, 99+%
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 99+%

CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 270102500
Price (USD)
71.60
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
71.60
Each
Chemical Identifiers
CAS5470-11-1
IUPAC Namehydroxylamine hydrochloride
Molecular FormulaClH4NO
InChI KeyWTDHULULXKLSOZ-UHFFFAOYSA-N
SMILESCl.NO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration with NaOH99.0 to 101.0 %
Sulfate (SO4)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only