Fehling B solution
Fehling B solution
Fehling B solution
Fehling B solution
Thermo Scientific Chemicals

Fehling B solution

CAS: 6381-59-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 270790025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
Boiling Point100.0°C
CAS6381-59-5
CAS1310-73-2
CAS7732-18-5
Chemical Name or MaterialFehling B solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only