2-Acetamido-2-deoxy-D-glucopyranose, 98%
2-Acetamido-2-deoxy-D-glucopyranose, 98%
2-Acetamido-2-deoxy-D-glucopyranose, 98%
Thermo Scientific Chemicals

2-Acetamido-2-deoxy-D-glucopyranose, 98%

Have Questions?
Catalog NumberQuantityPackaging
27094010010 gGlass bottle
Catalog number 270940100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7512-17-6
IUPAC NameN-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular FormulaC8H15NO6
InChI KeyOVRNDRQMDRJTHS-WZWLWIOANA-N
SMILESCC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific optical rotation+39° to +45° (20°C, 589 nm) (c=2, water, after 2 h)
HPLC>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only