3-Amino-4-hydroxybenzoic acid, 98%
3-Amino-4-hydroxybenzoic acid, 98%
3-Amino-4-hydroxybenzoic acid, 98%
Thermo Scientific Chemicals

3-Amino-4-hydroxybenzoic acid, 98%

CAS: 1571-72-8 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 271490250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1571-72-8
IUPAC Name3-amino-4-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyMRBKRZAPGUCWOS-UHFFFAOYSA-N
SMILESNC1=CC(=CC=C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to grey to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Melting point198°C to 205°C (Dec)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only