SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks

SoftFit-L™ Filtered Pipette Tips in Hinged Racks

Pipette samples with the accuracy and precision you expect from your pipette, every time with these pipette tips. For use with Rainin™ LTS pipettes.

Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)Tamanho da unidade
2739-HR300 μLCase of 4800
2779-HR1000 μLCase of 3072
2779-HRPK
conhecido também como 2779-HR
1000 μLPack of 768
2749-HR20 μLCase of 4800
2749-HRPK
conhecido também como 2749-HR
20 μLPack of 960
2739-HRPK
conhecido também como 2739-HR
300 μLPack of 960
2769-HR200 μLCase of 4800
2769-HRPK
conhecido também como 2769-HR
200 μLPack of 960
2789-HR1200 μLCase of 3072
2789-HRPK
conhecido também como 2789-HR
1200 μLPack of 768
Número do catálogo 2739-HR
Preço (BRL)
-
Volume (Metric):
300 μL
Unit Size:
Case of 4800
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Especificações
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description300 μL, Filtered, Sterile, Hinged Rack
Length (English)2.26 in.
Length (Metric)5.75cm
Model300L
ColorNatural
Color CodeOrange
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)300 μL
Unit SizeCase of 4800