2-Hydroxy-gamma-butyrolactone, 99%
2-Hydroxy-gamma-butyrolactone, 99%
2-Hydroxy-gamma-butyrolactone, 99%
Thermo Scientific Chemicals

2-Hydroxy-gamma-butyrolactone, 99%

CAS: 19444-84-9 | C4H6O3 | 102.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2750700505 gGlass bottle
Catalog number 275070050
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS19444-84-9
IUPAC Name3-hydroxyoxolan-2-one
Molecular FormulaC4H6O3
InChI KeyFWIBCWKHNZBDLS-UHFFFAOYNA-N
SMILESOC1CCOC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
Appearance (Form)Viscous liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only