Diaza-18-crown-6, 98%
Diaza-18-crown-6, 98%
Diaza-18-crown-6, 98%
Thermo Scientific Chemicals

Diaza-18-crown-6, 98%

CAS: 23978-55-4 | C12H28N2O4 | 264.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 275820025
Price (EUR)
275,00
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
275,00
Each
Chemical Identifiers
CAS23978-55-4
IUPAC Name1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium
Molecular FormulaC12H28N2O4
InChI KeyNLMDJJTUQPXZFG-UHFFFAOYSA-P
SMILESC1COCCOCC[NH2+]CCOCCOCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Glistening crystalline powder
Infrared spectrumConforms
Melting point111°C to 115°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only