SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks
SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks
SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Thermo Scientific™

SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks

The pipette tip and rack mail-back program provides a simple and effective solution for recycling PP#5 single use plastics. The TipCycle Kit included with your case of pipette tips to collect used pipette tips and empty racks. Complete waiver, attach pre-paid shipping label and return for recycling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
2769RC-HR200 μL
2739RC-HR300 μL
2749RC-HR20 μL
2779RC-HR1000 μL
2789RC-HR1200 μL
Catalog number 2769RC-HR
Price (USD)
-
Volume (Metric):
200 μL

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Stackable hinged racks save space in your lab
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Lid rim prevents racks from sliding around when stacked and carted around a lab
Specifications
Certifications/ComplianceDNase, RNase, DNA, ATP, and Endotoxin Free
Clean ClaimsNatural
For Use With (Application)Rainin™ LTS™ LiteTouch™
Length (English)1.97 in.
Length (Metric)5.00 cm
Model200 μL, Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
AutoclavableNot Autoclavable
ColorGreen
FilteredFiltered
FormatHinged Rack
Product LineSoftfit-L
SterilitySterile
Volume (Metric)200 μL
Unit SizeCase of 4800