Can you provide the shelf-life for Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent)?
Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.