Search
View additional product information for 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 - FAQs (277143)
1 product FAQs found
Yes. The Magnetic Stand-96 (Cat. No. AM10027) is compatible with 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 (Cat. No. 277143).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.