Specifications
Dimensions (L x W x H)128 x 86 x 41 mm
FormatStandard
LidWithout Lid
No. per Case32
No. per Pack4
Packaging32 Cases, 4 Pk.
Product TypeMicroplate
Quantity32/Cs.
Volume (Metric) Working900 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentUntreated
Well ShapeRound Bottom (U-well)
Unit SizeCase of 32