trans-2-Pentenoic acid, 97%
trans-2-Pentenoic acid, 97%
trans-2-Pentenoic acid, 97%
Thermo Scientific Chemicals

trans-2-Pentenoic acid, 97%

CAS: 13991-37-2 | C5H8O2 | 100.12 g/mol
Have Questions?
Catalog NumberQuantity
27937025025 g
Catalog number 279370250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13991-37-2
IUPAC Name(2E)-pent-2-enoic acid
Molecular FormulaC5H8O2
InChI KeyYIYBQIKDCADOSF-ONEGZZNKSA-N
SMILESCC\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4505 to 1.4525 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only