Search
View additional product information for RBS™ Solid Detergent - FAQs (27968)
1 product FAQs found
Only RBS Solid (Cat. No. 27968) is a non-foaming detergent that is suitable for automatic washers.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.