Product Image
Thermo Scientific™

Additional Dispenser Kit for Fluoroskan Ascent™ FL Microplate Fluorometer and Luminometer

Perform a number of fluorometric and luminometric research applications with the Thermo Scientific™ Fluoroskan Ascent™ FL Microplate Fluorometer and Luminometer.
Have Questions?
Catalog NumberFor Use With (Equipment)
2805630Fluoroskan
Catalog number 2805630
Price (USD)
-
For Use With (Equipment):
Fluoroskan
Perform a number of fluorometric and luminometric research applications with the Thermo Scientific™ Fluoroskan Ascent™ FL Microplate Fluorometer and Luminometer, which offers versatile plate formats, fast plate reading speeds, top/bottom plate reading and up to three dispensers. Compact and robust, the excellent optical performance of this machine also allows it to perform ideally on life science research applications.

Fast reading speed:

  • Reads a 96-well plate in just 15 seconds, which is essential for kinetic applications.
  • Combination of fluorometric GFP measurement and a lumometric luciferase measurement from the same well is fast due to the versatility of assay programming.

Onboard dispensers for exact follow-up of kinetic reactions:

  • Equipped with up to three reagent dispensers for rapid kinetic assays, such as Ca2+ flux
  • Supports simultaneous dispensing and reading, enabling monitoring of kinetic measurements from the very start of the reaction
  • Very low dead volume and the backflush capability for conservation of expensive reagent

High sensitivity for both top and bottom reading:

  • Fiberless direct illumination optics for both top and bottom reading
  • High sensitivity, wide dynamic range and low cross talk for accurate and precise results for 1- to 384-well plates

Specially designed for automation:

  • Reads up to 384-well plates for increased assay throughput
  • Easily integrated with automated systems

Includes:

  • PC Software and filter pairs:
    • Excitation: 355 nm, Emission: 460 nm
    • Excitation: 485 nm, Emission: 538 nm
    • Other filters available upon request

Recommended for:

Ca2+ flux assays, Cell proliferation, Cytotoxicity, Cell adhesion, DNA quantitation, Reporter gene assays, Hybridization assays, Ion Channel, Quantitation of PCR products, FRET assays, BRET assays, Molecular beacon assays, Immunoassays, Enzyme activity, Bacterial quantitation, Phagocytosis, Oligonucleotide assays

Specifications
Product LineFluoroskan Ascent
For Use With (Equipment)Fluoroskan
Product TypeAdditional Dispenser Kit
Unit SizeEach