Pierce™ 20X Phosphate Buffered Saline, 500 mL - FAQs
Pierce™ 20X Phosphate Buffered Saline, 500 mL - FAQs
View additional product information for Pierce™ Concentrated Buffer Stocks (10X and 20X) - FAQs (28352, 28341, 28363, 28362, 28346, 28354, 28344, 28355, 28358, 28348, 28360)
2 product FAQs found
What is the optimal pH for the Pierce Concentrated Buffer Stocks (10X and 20X) (Cat. No. 28362)?
The quality specifications for the ready to use 1x solution is 8.5 ± 0.2. (see example COA: https://assets.thermofisher.com/TFS-Assets/LSG/certificate/Certificates-of-Analysis/28362_WH332690.PDF). The pH should be between 8.3 and 8.7.
Find additional tips, troubleshooting help, and resources within our
Protein Purification and Isolation Support Center.
Can you provide the shelf-life for the Pierce 20X Phosphate Buffered Saline?
The Pierce 20X Phosphate Buffered Saline is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our
Protein Purification and Isolation Support Center.