Product Image
Gibco™

Tryptic Soy Broth, without dextrose

Tryptic Soy Broth without Dextrose is an enzymatic digest of casein and soybean meal. It is a low carbohydrate basalRead more
Have Questions?
Catalog NumberQuantity
28621010 kg
Catalog number 286210
Price (MXN)
23,952.55
Each
Quantity:
10 kg
Price (MXN)
23,952.55
Each
Tryptic Soy Broth without Dextrose is an enzymatic digest of casein and soybean meal. It is a low carbohydrate basal medium used for cultivating fastidious and nonfastidious microorganisms. Dextrose is omitted from the formula to permit use of the medium in fermentation studies where carbohydrates may be added. The carbohydrate concentration used most frequently in fermentation reactions is 0.5% or 1%.
Specifications
For Use With (Application)Microbial Fermentation
Media TypeDehydrated Culture Media
Packaging TypePail
Product LineGibco, Bacto
Quantity10 kg
Regulatory StatusFor Research Use or Further Manufacturing. Not for diagnostic use or direct administration in to humans or animals
Shelf Life5 Years
Solubility2.75% solution, soluble in distilled or deionized water.
FormPowder
pH7.1 to 7.5
Unit SizeEach