Ursolic acid, 95%
Ursolic acid, 95%
Ursolic acid, 95%
Thermo Scientific Chemicals

Ursolic acid, 95%

CAS: 77-52-1 | C30H48O3 | 456.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 290180010
Price (USD)
372.65
Online exclusive
414.00
Save 41.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
372.65
Online exclusive
414.00
Save 41.35
Each
Chemical Identifiers
CAS77-52-1
IUPAC Name(1S,2R,4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Molecular FormulaC30H48O3
InChI KeyWCGUUGGRBIKTOS-GPOJBZKASA-N
SMILESC[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=94 %
Specific optical rotation+57° to +61° (20°C, 589 nm) (c=0.3, pyridine)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only