503 Service Temporarily Unavailable
503 Service Temporarily Unavailable
nginx
Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Actual product may vary
Thermo Scientific™
Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
290199Sensor
400159Replacement Back-Up Battery
Catalog number 290199
Price (MXN)
-
Type:
Sensor
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Specifications
DefrostAutomatic
DescriptionOptional PT100 sensor for connection to building automation systems (Factory installed)
For Use With (Equipment)Revco™ ExF/DxF, Forma™ 900, HERAfreeze™ HFU-B, TSE/TSD Series
TypeSensor
Unit SizeEach
503 Service Temporarily Unavailable
503 Service Temporarily Unavailable
nginx