4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
Thermo Scientific Chemicals

4-tert-Butylcalix[4]arene, 99%

CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol
製品番号(カタログ番号) 290220250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS60705-62-6
IUPAC Name5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular FormulaC44H56O4
InChI KeyNVKLTRSBZLYZHK-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
さらに表示
Appearance (Form)Crystalline powder or crystals or flakes
Appearance (Color)White to light beige
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only