Mini Capillary Unions
Actual product may vary
Thermo Scientific™
Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
290GU4980.22 to 0.32 mm ID Capillary GC Column
290GU4990.53 mm ID Capillary GC Column
Catalog number 290GU498
Price (USD)
88.20
Each
For Use With (Equipment):
0.22 to 0.32 mm ID Capillary GC Column
Price (USD)
88.20
Each
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
Specifications
DescriptionFor 0.32 mm I.D. Capillary
For Use With (Equipment)0.22 to 0.32 mm ID Capillary GC Column
TypeCapillary Union
Unit SizeEach