SilTite™ Ferrule Kits
Actual product may vary
Thermo Scientific™
SilTite™ Ferrule Kits
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Diameter (Metric) Inside
290MA195Thermo Scientific ISQ GC-MS Interface0.32 mm
290MA194Thermo Scientific ISQ GC-MS Interface0.1 to 0.25 mm
290MA196Thermo Scientific ISQ GC-MS Interface0.53 mm
290MA215Thermo Scientific TRACE 1300/1310 GC SSL Injectors0.1 to 0.25 mm
290MA216Thermo Scientific TRACE 1300/1310 GC SSL Injectors0.32 mm
290MA217Thermo Scientific TRACE 1300/1310 GC SSL Injectors0.53 mm
290MT229Thermo Scientific DSQ GC-MS Interface0.1 to 0.25 mm
290MT230Thermo Scientific DSQ GC-MS Interface0.32 mm
290MT231Thermo Scientific DSQ GC-MS Interface0.53 mm
Catalog number 290MA195
Price (USD)
-
For Use With (Equipment):
Thermo Scientific ISQ GC-MS Interface
Diameter (Metric) Inside:
0.32 mm
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description290 MA195
Diameter (Metric) Inside0.32 mm
Includes2 SilTite Nuts, 10 Ferrules
For Use With (Equipment)Thermo Scientific ISQ GC-MS Interface
MaterialSilTite Metal
TypeFerrule Kit
Unit SizePack of 10