See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Chromatography Supplies
▸
Chromatography Ferrule and Nut Kits
▸
Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Actual product may vary
Thermo Scientific™
Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Thermo Scientific Finger Tite Connectors for Thermo Scientific and Agilent Instruments offer easier fit, reliability and leak-free connections.
Have Questions?
Change view
Catalog Number
Type
290SA133
Split/Splitless Injector Base Seal
290SA131
Split/Splitless and MS Starter Kit
290SA132
Split/Splitless and FID Starter Kit
3 Options
Catalog number
290SA133
Price (USD)
-
Request A Quote
Type:
Split/Splitless Injector Base Seal
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
Easy to handle with the nut touchable even with a hot injector/detector
No tools required
Specifications
Description
Injector Base Seal; 2/Pk.
Quantity
2/Pk.
For Use With (Equipment)
TRACE 1300 Series GC and Agilent GC Instruments
Type
Split/Splitless Injector Base Seal