(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 291632500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2041-14-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only