2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
Thermo Scientific Chemicals

2-Chloro-1H-benzimidazole, 97%

CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Catalog NumberQuantity
29196025025 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 291960250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4857-06-1
IUPAC Name2-chloro-1H-1,3-benzodiazole
Molecular FormulaC7H5ClN2
InChI KeyAYPSHJCKSDNETA-UHFFFAOYSA-N
SMILESClC1=NC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=96.0 %
Appearance (Color)Beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only