Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium
Thermo Scientific Chemicals

Bis(dibenzylideneacetone)palladium

CAS: 32005-36-0 | C34H28O2Pd | 575.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 291970010
Price (USD)
79.65
Online exclusive
88.10
Save 8.45
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.65
Online exclusive
88.10
Save 8.45
Each
Chemical Identifiers
CAS32005-36-0
IUPAC Namebis((1E,4E)-1,5-diphenylpenta-1,4-dien-3-one) palladium
Molecular FormulaC34H28O2Pd
InChI KeyUKSZBOKPHAQOMP-SVLSSHOZSA-N
SMILES[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Red-brown to black
Assay16 to 21 % (Pd)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only