Pierce™ NeutrAvidin™ Agarose, 10 mL - FAQs
View additional product information for Pierce™ NeutrAvidin™ Agarose - FAQs (29200, 29201)
1 product FAQs found
I received Pierce NeutrAvidin Agarose (Cat. No. 29200) at ambient temperature, even though it is supposed to be stored at -20 degrees C. Is the performance of the product still reliable?
Yes, Pierce NeutrAvidin Agarose is routinely shipped at room temperature. This product is stable at ambient temperatures for short periods of time.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.