Difco™ TC Yeastolate, ultra-filtered (UF)
Gibco™

Difco™ TC Yeastolate, ultra-filtered (UF)

Enhance cell culture performance with Gibco™ Difco™ TC Yeastolate Ultra-Filtered (UF) Low Bioburden while also mitigating risk in bioprocessing methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityManufacturing Quality
29280510 kgUF
292804500 gUF
67007950 kgUF
A80501-0110 kgUF, Low Bioburden (≤100 CFU)
A80501-0250 kgUF, Low Bioburden (≤100 CFU)
A80503-0110 kgUF, Low Bioburden (≤300 CFU)
A80503-0250 kgUF, Low Bioburden (≤300 CFU)
Catalog number 292805
Price (USD)
6,390.00
Each
Quantity:
10 kg
Manufacturing Quality:
UF
Price (USD)
6,390.00
Each
Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.

Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.

Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU

Specifications
ColorBeige
Culture TypeMammalian, Insect, Microbial
Endotoxin Level500 EU/g
For Use With (Application)Preparing bacterial, insect, and mammalian cell culture media
FormatPail
Manufacturing QualityUF
Product LineDifco
Quantity10 kg
FormPowder
Product TypeTC Yeastolate
pH5.4 to 7.2
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.