Methacrolein, 90%, stabilized
Methacrolein, 90%, stabilized
Methacrolein, 90%, stabilized
Thermo Scientific Chemicals

Methacrolein, 90%, stabilized

Methacrolein, 90%, C4H6O, CAS Number-78-85-3, 2-methyl-2-propenal, 2-methylpropenal, 2-methylacrolein, 2-methylenepropanal, 2-propenal, 2-methyl, isobutenal, methacrylaldehyde, methacrylic aldehyde, methylacrylaldehyde, methacrolein, 50mL, 88% min. (GC), 01, III, 2981, 69.0 deg.C | CAS: 78-85-3 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 292830100
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Chemical Identifiers
CAS78-85-3
IUPAC Name2-methylprop-2-enal
Molecular FormulaC4H6O
InChI KeySTNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILESCC(=C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Stabilizer0.1 to 0.5 % Hydroquinone
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only