Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
Thermo Scientific Chemicals

Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1

CAS: 4137-10-4 | C5H14N2O2 | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 292905000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4137-10-4
SpecificationsSpecification SheetSpecification Sheet
Ratio< 2.0/1 Carbon dioxide/Dimethylamine
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Water=<1 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only