11-Bromo-1-undecanol, 97%
11-Bromo-1-undecanol, 97%
11-Bromo-1-undecanol, 97%
Thermo Scientific Chemicals

11-Bromo-1-undecanol, 97%

CAS: 1611-56-9 | C11H23BrO | 251.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 293440500
Price (USD)
176.65
Special offer
Online exclusive
Ends: 28-Dec-2026
208.00
Save 31.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
176.65
Special offer
Online exclusive
Ends: 28-Dec-2026
208.00
Save 31.35
Each
Chemical Identifiers
CAS1611-56-9
IUPAC Name11-bromoundecan-1-ol
Molecular FormulaC11H23BrO
InChI KeyXFGANBYCJWQYBI-UHFFFAOYSA-N
SMILESOCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point42°C to 47°C
Appearance (Color)White to beige
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only