4-(Trifluoromethoxy)benzaldehyde, 99%
4-(Trifluoromethoxy)benzaldehyde, 99%
4-(Trifluoromethoxy)benzaldehyde, 99%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzaldehyde, 99%

CAS: 659-28-9 | C8H5F3O2 | 190.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 293500100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS659-28-9
IUPAC Name4-(trifluoromethoxy)benzaldehyde
Molecular FormulaC8H5F3O2
InChI KeyXQNVDQZWOBPLQZ-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow-green
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only