Bis(diphenylphosphino)methane, 98%
Bis(diphenylphosphino)methane, 98%
Bis(diphenylphosphino)methane, 98%
Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 98%

CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 293610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2071-20-7
IUPAC Name[(diphenylphosphanyl)methyl]diphenylphosphane
Molecular FormulaC25H22P2
InChI KeyXGCDBGRZEKYHNV-UHFFFAOYSA-N
SMILESC(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=97.5 %
Melting point118°C to 122°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only