4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
Thermo Scientific Chemicals

4-Methoxybenzylchloride, 98%, stabilized

CAS: 824-94-2 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 294170250
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Chemical Identifiers
CAS824-94-2
IUPAC Name1-(chloromethyl)-4-methoxybenzene
Molecular FormulaC8H9ClO
InChI KeyMOHYOXXOKFQHDC-UHFFFAOYSA-N
SMILESCOC1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
Stabilizer1 to 5 % K2CO3
GC>=97.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only