CaptureSelect™ Lenti VSVG Affinity Matrix
Thermo Scientific™

CaptureSelect™ Lenti VSVG Affinity Matrix

Thermo Scientific™ CaptureSelect™ Lenti VSVG Affinity Matrix has been designed specifically for the purification of VSV-G pseudotyped lentivirus particles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294393205050 mL
29439320055 mL
294393201010 mL
Catalog number 2943932050
Price (USD)
8,160.00
Each
Quantity:
50 mL
Price (USD)
8,160.00
Each

CaptureSelect Lenti VSVG Affinity Matrix has been designed specifically for the purification of VSV-G pseudotyped lentivirus particles.

Features of this affinity matrix include:

  • Efficient and high-yield single-step purification of lentivirus particles from recombinant suspension cultivated harvest
  • Gentle, neutral pH elution conditions to retain lentiviral activity
  • Excellent scalability
  • Non-animal derived

CaptureSelect Lenti VSVG Affinity Matrix offers an efficient and scalable purification method for VSV-G pseudotyped lentivirus particles, with a binding capacity of 1 x 1011 total particles/mL resin. Elution can be performed using gentle elution conditions, which helps to retain infectivity of the enveloped virus particles.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect Lenti VSVG affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: ∼1 x 1011 total particles/mL matrix
  • Elution conditions: 50 mM HEPES, 150 mM NaCl, 0.8 M arginine, pH 7.5
  • Strip conditions: 50 mM sodium phosphate, pH 12
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity∼1 x 1011 total particles/mL matrix
Flow Rate50 to 200 cm/hr. (up to 2 bar)
For Use With (Application)Lenti VSV-G particle purification
Formulation20% (v/v) ethanol
Ligand TypeCaptureSelect Lenti VSV-G Affinity Ligand
MatrixAgarose-based, epoxide activated
Quantity50 mL
TargetLenti virus VSV-G envelop glycoprotein
Capacity (Metric)∼1 x 1011 total particles/mL matrix
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light