CaptureSelect™ IgA-XL Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgA-XL Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgA-XL Affinity Matrix is intended for the purification of human IgA, with no binding to other immunoglobulins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2943972500500 mL
29439720055 mL
294397201010 mL
294397205050 mL
Catalog number 2943972500
Price (USD)
25,970.00
Each
Quantity:
500 mL
Price (USD)
25,970.00
Each

CaptureSelect IgA-XL Affinity Matrix is intended for the purification of human IgA, with no binding to other immunoglobulins. The IgA-XL affinity matrix provides improved binding capacities (>15 g/L IgA) and elution characteristics (full elution at pH 4) compared to CaptureSelect IgA Affinity Matrix.

Features of this affinity matrix include:

  • Purification of all IgA serotypes: IgA1, IgA2, dimeric IgA, and S-IgA (secretory IgA)
  • IgA purification from plasma, serum, and recombinant sources
  • Efficient purification of IgA with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

CaptureSelect IgA-XL Affinity Matrix binds to the Fc part of IgA. The high selectivity and yields obtained using IgA-XL Affinity Matrix enable a robust, fast, and efficient purification process with excellent purity obtained in one step.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect IgA-XL affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: > 15 g/L at 5 minutes residence time, tested with plasma-derived polyclonal IgA
  • Elution conditions: 20 mM sodium citrate, pH 4
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures
Specifications
Ligand TypeCaptureSelect IgA-XL Affinity Ligand
MatrixAgarose
Max. Operating Pressure2 bar
Quantity500 mL
Shipping ConditionWet Ice
Capacity (Metric)>15 g IgA/L
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in a light-protected environment. Avoid direct exposure to sunlight. Do not freeze