Search
Search

| Catalog Number | Quantity |
|---|---|
| 2944062005 | 5 mL |
| 2944062010 | 10 mL |
| 2944062050 | 50 mL |
CaptureSelect Factor H Affinity Matrix is designed for the purification of human complement Factor H from plasma and recombinant sources.
Features of the CaptureSelect Factor H Affinity Matrix include:
The high selectivity and yields obtained using CaptureSelect Factor H Affinity Matrix enable a robust, fast, and efficient purification process with excellent purity in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created using a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
Matrix: agarose-based
Average particle size: 65 ± 10 μm
Ligand: CaptureSelect Factor H affinity ligand
Ligand coupling method: epoxide
Binding capacity: > 2 mg Factor H/mL resin
Binding conditions: PBS or TBS, pH 7
Elution conditions: 20 mM citric acid, pH 3
Flow characteristics: 50–200 cm/h (up to 2 bar)
Formulation buffer: 20% (v/v) ethanol